Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sll
Entrez Gene ID
42115
Gene Name
slalom
Gene Aliases
1422/14, CG7623, Dmel\CG7623, Dmel_CG7623, Sll, anon-WO0118547.555, anon-WO0172774.85, l(3)S142214
Chromosome Location
Chr.3R: 17606106 - 17608541 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038524
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260224.2 NP_001247153.1 3 - 4 1472 64
NM_079665.4 NP_524389.1 3 - 4 1465 64
GenBank mRNA AB107958.1 - 2 - 3 1113 64
BT132969.1 - 3 - 4 1395 64

View Full Product Details