Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AttD
- Entrez Gene ID
-
42122
- Gene Name
- Attacin-D
- Gene Aliases
-
Att, Attacin D, CG7629, Dmel\CG7629, Dmel_CG7629, att, att D, attD
- Chromosome Location
-
Chr.3R: 17625268 - 17626086 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038530
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079667.3 |
NP_524391.2 |
1 - 2 |
212 |
65 |
View Full Product Details