Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rh2
Entrez Gene ID
42261
Gene Name
Rhodopsin 2
Gene Aliases
CG16740, DMELRH2, Dm Rh2, Dmel\CG16740, Dmel_CG16740, RH2, Rh, rh2
Chromosome Location
Chr.3R: 18899423 - 18901019 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003248
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079674.3 NP_524398.1 2 - 3 1068 75
GenBank mRNA BT150161.1 - 2 - 3 1069 75

View Full Product Details