Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- cdi
- Entrez Gene ID
-
42289
- Gene Name
- center divider
- Gene Aliases
-
91F, B9.3, CDI/TSKI, CG6027, Cdi, Cdi/TESK1, Dmel\CG6027, Dmel_CG6027, cdi/TESK1, l(3)07013
- Chromosome Location
-
Chr.3R: 19044882 - 19094270 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004876
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079677.4 |
NP_524401.2 |
5 - 6 |
1884 |
63 |
GenBank mRNA |
BT004492.1 |
- |
4 - 5 |
930 |
63 |
View Full Product Details