Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dys
Entrez Gene ID
42327
Gene Name
Dystrophin
Gene Aliases
CG17750, CG31175, CG34157, CG7240, CG7243, CG7344, DLP, DLP1, DLP186, DLP2, DLP3, DYS, DmDLP, DmDYS, Dmel\CG34157, Dmel_CG34157, Dp117, Dp186, Dp205, GI3046716, IDLP, det, dmDLP, dmDp186, dmDys, dys
Chromosome Location
Chr.3R: 19461085 - 19597288 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260003
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043257.2 NP_001036722.1 10 - 11 6555 71
NM_001043258.2 NP_001036723.1 10 - 11 6555 71
NM_001043259.2 NP_001036724.1 9 - 10 5556 71
NM_001043261.2 NP_001036726.1 9 - 10 6268 71
NM_001043263.2 NP_001036728.1 9 - 10 6482 71
NM_001275801.1 NP_001262730.1 8 - 9 5441 71
NM_001275803.1 NP_001262732.1 9 - 10 5556 71
GenBank mRNA AF277386.1 - 8 - 9 5125 71

View Full Product Details