Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hs6st
Entrez Gene ID
42380
Gene Name
Heparan sulfate 6-O-sulfotransferase
Gene Aliases
CG11683, CG17188, CG18533, CG4451, DmHs6st, Dmel\CG4451, Dmel_CG4451, HS6ST, dHS6ST, hs6st
Chromosome Location
Chr.3R: 19929277 - 20008512 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038755
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275821.1 NP_001262750.1 3 - 4 1111 72
NM_079685.3 NP_524409.2 3 - 4 1111 72
GenBank mRNA AY051902.1 - 3 - 4 1088 72

View Full Product Details