Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dhc93AB
Entrez Gene ID
42485
Gene Name
Dynein heavy chain at 93AB
Gene Aliases
CG3723, DHC, DHC4C, Dhc, Dhc 93AB, Dmel\CG3723, Dmel_CG3723, dhc, dhc93AB
Chromosome Location
Chr.3R: 21024580 - 21042499 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013812
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275864.1 NP_001262793.1 28 - 29 13407 67
NM_079700.3 NP_524424.2 27 - 28 13377 67

View Full Product Details