Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slmb
Entrez Gene ID
42504
Gene Name
supernumerary limbs
Gene Aliases
BcDNA:GM02031, CG3412, Dmel\CG3412, Dmel_CG3412, FBXW1, MENE (3R)-B, MENE(3R)-B, SLIMB, SLMB, Slimb, Slimb/Beta-TRCP1, Slimb/beta-TRCP, Slimb/beta-TrCP, Slimb/betaTrCP, Slmb, beta-TrCP, betaTrCP, crd, ica, l(3)00295, shv, slimb, wel
Chromosome Location
Chr.3R: 21119252 - 21127485 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0283468
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275872.1 NP_001262801.1 3 - 4 1045 61
NM_001275873.1 NP_001262802.1 3 - 4 1045 61
NM_079706.3 NP_524430.1 3 - 4 1045 61
GenBank mRNA AF222923.1 - 3 - 4 1042 61
AF222924.1 - 3 - 4 924 61
AY118898.1 - 3 - 4 1013 61

View Full Product Details