Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- tin
- Entrez Gene ID
-
42536
- Gene Name
- tinman
- Gene Aliases
-
CG7895, DROHOXHK4, DROHOXNK4, DmNK-4, Dmel\CG7895, Dmel_CG7895, HOX, NK-4, NK-4/msh-2, NK4, NK4/msh-2, NKX2.5, TIN, Tin, msh-2, msh2, tin/NK4
- Chromosome Location
-
Chr.3R: 21378977 - 21381970 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004110
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079709.3 |
NP_524433.1 |
2 - 3 |
1067 |
106 |
GenBank mRNA |
BT050510.1 |
- |
2 - 3 |
1065 |
106 |
View Full Product Details