Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GABA-B-R2
Entrez Gene ID
42561
Gene Name
metabotropic GABA-B receptor subtype 2
Gene Aliases
BcDNA:GH07312, CG6706, D-GABA[[B]]R2, D-Gaba2, Dmel\CG6706, Dmel_CG6706, GABA B R2, GABA-BR2, GABA-[[B]]R2, GABABR-R2, GABABR2, GABA[[B]]-R2, GABA[[B]]R2, GB2, GBR, GH07312, anon-WO0170980.58, anon-WO0170980.59
Chromosome Location
Chr.3R: 21770333 - 21775840 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027575
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079714.2 NP_524438.1 9 - 10 2245 83
NM_169968.1 NP_732658.1 9 - 10 2245 83
GenBank mRNA AF145639.1 - 9 - 10 2239 83

View Full Product Details