Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CenB1A
Entrez Gene ID
42735
Gene Name
Centaurin beta 1A
Gene Aliases
ACAP, CG6742, Cen1ba, Dmel\CG6742, Dmel_CG6742, cenB1A
Chromosome Location
Chr.3R: 23118400 - 23124217 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039056
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079734.5 NP_524458.1 9 - 10 2691 64
NM_170049.4 NP_732826.2 5 - 6 1792 64
GenBank mRNA AF254071.1 - 9 - 10 2302 64
BT122085.1 - 9 - 10 2630 64

View Full Product Details