Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vps24
Entrez Gene ID
40542
Gene Name
Vacuolar protein sorting 24
Gene Aliases
CG9779, DmVPS24, Dmel\CG9779, Dmel_CG9779, ESCRT, dvps24, vps24
Chromosome Location
Chr.3R: 4307435 - 4310947 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037231
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316528.1 NP_001303457.1 2 - 3 396 75
NM_141194.3 NP_649451.1 2 - 3 396 75
GenBank mRNA AY058381.1 - 2 - 3 357 75

View Full Product Details