Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Syt14
Entrez Gene ID
40544
Gene Name
Synaptotagmin 14
Gene Aliases
CG9778, Dmel\CG9778, Dmel_CG9778, Syt 14, Syt16, dsyt14, syt14, syt16
Chromosome Location
Chr.3R: 4314455 - 4319069 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261086
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275320.1 NP_001262249.1 3 - 4 981 110
NM_001275321.1 NP_001262250.1 3 - 4 981 110
NM_141196.2 NP_649453.1 3 - 4 981 110
GenBank mRNA AY122072.1 - 3 - 4 650 110

View Full Product Details