Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Skp2
Entrez Gene ID
40548
Gene Name
S-phase kinase-associated protein 2
Gene Aliases
153298_at, CG9772, Dmel\CG9772, Dmel_CG9772, F-box, FBXL1, SKP2, dSkp2
Chromosome Location
Chr.3R: 4334879 - 4337674 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037236
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_164340.4 NP_730815.2 1 - 2 127 81
NM_164341.4 NP_730816.3 1 - 2 127 81

View Full Product Details