Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- lost
- Entrez Gene ID
-
40559
- Gene Name
- CG14648-PA
- Gene Aliases
-
CG14648, Dmel\CG14648, Dmel_CG14648, Growl, anon-WO0118547.126, growl, nev, nvd
- Chromosome Location
-
Chr.3R: 4403076 - 4407182 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0263594
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141209.3 |
NP_649466.1 |
5 - 6 |
1377 |
75 |
GenBank mRNA |
AY058608.1 |
- |
5 - 6 |
1352 |
75 |
View Full Product Details