Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Prosbeta2R2
- Entrez Gene ID
-
40621
- Gene Name
- Proteasome beta2 subunit-related 2
- Gene Aliases
-
CG12161, Dmel\CG12161, Dmel_CG12161, beta2R2
- Chromosome Location
-
Chr.3R: 5217751 - 5219528 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037296
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141258.4 |
NP_649515.3 |
2 - 3 |
361 |
69 |
GenBank mRNA |
AY075170.1 |
- |
2 - 3 |
279 |
69 |
View Full Product Details