Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mRpL44
- Entrez Gene ID
-
40656
- Gene Name
- mitochondrial ribosomal protein L44
- Gene Aliases
-
CG2109, Dmel\CG2109, Dmel_CG2109, MRP-L44
- Chromosome Location
-
Chr.3R: 5469463 - 5470636 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037330
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141284.4 |
NP_649541.1 |
1 - 2 |
124 |
82 |
View Full Product Details