Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hcs
Entrez Gene ID
40659
Gene Name
Holocarboxylase synthetase
Gene Aliases
CG14670, Dmel\CG14670, Dmel_CG14670, HCS, HSC, NEST:bs27h05
Chromosome Location
Chr.3R: 5474899 - 5485393 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037332
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300246.1 NP_001287175.1 2 - 3 843 80
NM_141285.2 NP_649542.1 2 - 3 843 80
GenBank mRNA AY118365.1 - 2 - 3 840 80

View Full Product Details