Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Osi2
Entrez Gene ID
40756
Gene Name
Osiris 2
Gene Aliases
CG1148, Dmel\CG1148, Dmel_CG1148
Chromosome Location
Chr.3R: 6209373 - 6213493 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037410
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275374.1 NP_001262303.1 2 - 3 458 61
NM_141364.2 NP_649621.1 2 - 3 516 61
NM_169130.2 NP_731052.2 2 - 3 541 61
GenBank mRNA BT015251.1 - 2 - 3 430 61

View Full Product Details