Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Osi5
Entrez Gene ID
40759
Gene Name
Osiris 5
Gene Aliases
CG15590, Dmel\CG15590, Dmel_CG15590
Chromosome Location
Chr.3R: 6227512 - 6228828 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037413
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141367.3 NP_649624.1 1 - 2 513 81
GenBank mRNA BT028782.1 - 1 - 2 630 81

View Full Product Details