Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Osi6
Entrez Gene ID
40760
Gene Name
Osiris 6
Gene Aliases
BcDNA:LD21503, CG1151, Dmel\CG1151, Dmel_CG1151
Chromosome Location
Chr.3R: 6234708 - 6237181 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027527
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300263.1 NP_001287192.1 1 - 2 244 93
NM_141368.3 NP_649625.2 1 - 2 244 93
GenBank mRNA BT031257.1 - 1 - 2 222 93

View Full Product Details