Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Osi17
Entrez Gene ID
40774
Gene Name
Osiris 17
Gene Aliases
CG15598, Dmel\CG15598, Dmel_CG15598, mdcds_43703
Chromosome Location
Chr.3R: 6311772 - 6327308 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037427
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275377.1 NP_001262306.1 2 - 3 1962 81
NM_001275378.1 NP_001262307.1 1 - 2 1249 81
NM_001275379.1 NP_001262308.1 2 - 3 1962 81
NM_141381.3 NP_649638.2 1 - 2 1249 81
GenBank mRNA AY118624.1 - 1 - 2 1246 81

View Full Product Details