Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Osi19
Entrez Gene ID
40776
Gene Name
Osiris 19
Gene Aliases
CG15189, Dmel\CG15189, Dmel_CG15189, osi19
Chromosome Location
Chr.3R: 6335362 - 6337196 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037429
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170058.2 NP_001163529.1 3 - 4 714 64
NM_141383.2 NP_649640.1 3 - 4 741 64
GenBank mRNA BT015241.1 - 3 - 4 712 64

View Full Product Details