Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- pyd3
- Entrez Gene ID
-
40916
- Gene Name
- CG3027-PA
- Gene Aliases
-
CG3027, Dmel\CG3027, Dmel_CG3027, betaAS, betaASase
- Chromosome Location
-
Chr.3R: 7747913 - 7750079 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037513
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141475.3 |
NP_649732.1 |
1 - 2 |
195 |
93 |
GenBank mRNA |
AF333187.1 |
- |
1 - 2 |
151 |
93 |
View Full Product Details