Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CG7483
- Entrez Gene ID
-
40987
- Gene Name
- CG7483-PA
- Gene Aliases
-
7483, DmRH28, Dmel\CG7483, Dmel_CG7483, EIF4AIII, anon-WO0118547.364, eIF4A, eIF4A-III, eIF4A3, eIF4AIII, eIF4aIII, l(3)84Fi
- Chromosome Location
-
Chr.3R: 8313208 - 8314864 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037573
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141531.3 |
NP_649788.2 |
2 - 3 |
627 |
73 |
GenBank mRNA |
BT133445.1 |
- |
2 - 3 |
621 |
73 |
View Full Product Details