Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Vps16A
- Entrez Gene ID
-
41107
- Gene Name
- Vacuolar protein sorting 16A
- Gene Aliases
-
CG8454, DmVps16, Dmel\CG8454, Dmel_CG8454, V16, Vps16, Vps16a, dVps16A, ma, vps16A, vps16a
- Chromosome Location
-
Chr.3R: 9267019 - 9270224 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0285911
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141620.3 |
NP_649877.1 |
2 - 3 |
352 |
63 |
GenBank mRNA |
AY071611.1 |
- |
2 - 3 |
339 |
63 |
View Full Product Details