Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Task7
- Entrez Gene ID
-
41125
- Gene Name
- TWIK-related acid-sensitive K[+] channel 7
- Gene Aliases
-
CG9361, Dmel\CG9361, Dmel_CG9361, K2P-TASK7, TASK-7, dTASK-7
- Chromosome Location
-
Chr.3R: 9410329 - 9412293 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037690
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141634.2 |
NP_649891.1 |
2 - 3 |
561 |
70 |
GenBank mRNA |
BT024313.1 |
- |
2 - 3 |
562 |
70 |
View Full Product Details