Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- by
- Entrez Gene ID
-
41144
- Gene Name
- blistery
- Gene Aliases
-
3R-B, BcDNA:RE65113, CG9379, CT26639, Dmel\CG9379, Dmel_CG9379, tensin
- Chromosome Location
-
Chr.3R: 9528238 - 9533011 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000244
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141644.4 |
NP_649901.1 |
1 - 2 |
2271 |
82 |
GenBank mRNA |
AY094941.1 |
- |
1 - 2 |
2275 |
82 |
View Full Product Details