Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trbd
Entrez Gene ID
41179
Gene Name
trabid
Gene Aliases
CG9448, Dmel\CG9448, Dmel_CG9448, Q9VH90, Trbd
Chromosome Location
Chr.3R: 9705996 - 9711037 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037734
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141674.3 NP_649931.1 8 - 9 2851 64
GenBank mRNA AY058438.1 - 8 - 9 2813 64

View Full Product Details