Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Npc2d
- Entrez Gene ID
-
41232
- Gene Name
- Niemann-Pick type C-2d
- Gene Aliases
-
CG12813, Dmel\CG12813, Dmel_CG12813, npc2d
- Chromosome Location
-
Chr.3R: 10055356 - 10056083 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037782
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141718.2 |
NP_649975.1 |
1 - 2 |
100 |
58 |
View Full Product Details