Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
knk
Entrez Gene ID
47137
Gene Name
knickkopf
Gene Aliases
CG6217, Dmel\CG6217, Dmel_CG6217, Gnarled, Knk, dyspneic, gnar, l(3)85Eg
Chromosome Location
Chr.3R: 10122365 - 10125664 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001321
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141724.4 NP_649981.1 4 - 5 1661 86
GenBank mRNA AY071192.1 - 4 - 5 1659 86

View Full Product Details