Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAF1B
Entrez Gene ID
41242
Gene Name
TATA box-binding protein-associated factor RNA polymerase I subunit B
Gene Aliases
CG6241, Dmel\CG6241, Dmel_CG6241, TAF1b, Taf1B
Chromosome Location
Chr.3R: 10140103 - 10144184 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037792
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275521.1 NP_001262450.1 1 - 2 541 105
NM_001275522.1 NP_001262451.1 1 - 2 541 105
NM_141725.3 NP_649982.1 1 - 2 541 105
GenBank mRNA BT057993.1 - 1 - 2 531 105

View Full Product Details