Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- KP78b
- Entrez Gene ID
-
41361
- Gene Name
- CG17216-PA
- Gene Aliases
-
CG17216, Dmel\CG17216, Dmel_CG17216, KP78B, Kp78B, Kp78b
- Chromosome Location
-
Chr.3R: 11346294 - 11353131 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026063
- Gene Symbol
- KP78a
- Entrez Gene ID
-
41362
- Gene Name
- CG6715-PB
- Gene Aliases
-
CG6715, Dmel\CG6715, Dmel_CG6715, KP78A, Kp78A
- Chromosome Location
-
Chr.3R: 11346294 - 11353131 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026064
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141809.3 |
NP_650066.1 |
2 - 2 |
4830 |
99 |
View Full Product Details