Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dpr5
Entrez Gene ID
41381
Gene Name
defective proboscis extension response 5
Gene Aliases
CG5308, CT16895, Dmel\CG5308, Dmel_CG5308, Dpr-5, Dpr5
Chromosome Location
Chr.3R: 11499984 - 11505951 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037908
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141823.3 NP_650080.3 4 - 5 1206 67
NM_169395.2 NP_731580.1 3 - 4 808 67
GenBank mRNA AY047569.1 - 4 - 5 1206 67

View Full Product Details