Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Csk
Entrez Gene ID
41398
Gene Name
C-terminal Src kinase
Gene Aliases
CG17309, CG32852, CG42317, CSK, DCsk, Dmel\CG42317, Dmel_CG42317, Src, csk, d-Csk, dCSK, dCsk, dcsk, flik, l(3)S017909, l(3)j1D8
Chromosome Location
Chr.3R: 11633278 - 11650706 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262081
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275547.1 NP_001262476.1 5 - 6 2671 84
NM_141840.1 NP_650097.2 5 - 6 1927 84
NM_169414.2 NP_731607.1 5 - 6 2363 84
NM_169415.2 NP_731608.1 5 - 6 1791 84
NM_169416.2 NP_731609.1 4 - 5 2011 84
NM_169418.3 NP_731611.2 6 - 7 2932 84
NM_176461.1 NP_788638.1 5 - 6 1785 84

View Full Product Details