Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glo
Entrez Gene ID
41431
Gene Name
glorund
Gene Aliases
110/46, CG6946, Dmel\CG6946, Dmel_CG6946, Glo, anon-WO0118547.328, cg6946, l(3)S011046, l(3)S11046, p67, p67-K
Chromosome Location
Chr.3R: 11815206 - 11819339 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259139
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141863.3 NP_650120.1 4 - 5 796 61
NM_169434.2 NP_731639.1 4 - 5 721 61
GenBank mRNA AY089465.1 - 4 - 5 878 61

View Full Product Details