Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cyp9f2
- Entrez Gene ID
-
41520
- Gene Name
- CG11466-PA
- Gene Aliases
-
9f2, CG11466, CYP9F2, Cyp 9f2, Cyp9F2, Dmel\CG11466, Dmel_CG11466, anon-EST:Posey150
- Chromosome Location
-
Chr.3R: 12403340 - 12406028 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038037
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141932.3 |
NP_650189.1 |
1 - 2 |
316 |
85 |
GenBank mRNA |
AY058466.1 |
- |
1 - 2 |
224 |
85 |
View Full Product Details