Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
beat-Vc
Entrez Gene ID
41575
Gene Name
beaten path Vc
Gene Aliases
Beat-Vc, CG14390, Dmel\CG14390, Dmel_CG14390, beat Vc
Chromosome Location
Chr.3R: 12746036 - 12783216 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038084
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001369977.1 NP_001356887.1 5 - 6 1334 85
GenBank mRNA BT088886.1 - 4 - 5 911 85

View Full Product Details