Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
yrt
Entrez Gene ID
41656
Gene Name
yurt
Gene Aliases
CG9764, Dmel\CG9764, Dmel_CG9764, Nbl4, Yrt, Yurt, dlhB, l(3)87Ek, l(3)E19, l(3)m112, m112
Chromosome Location
Chr.3R: 13428867 - 13434984 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004049
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144573.2 NP_001138045.1 5 - 6 2459 108
NM_001144574.2 NP_001138046.1 5 - 6 2459 108
NM_142034.2 NP_650291.1 5 - 6 2459 108
GenBank mRNA BT015265.1 - 5 - 6 2449 108
EF078695.1 - 5 - 6 1747 108

View Full Product Details