Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
omd
Entrez Gene ID
41674
Gene Name
oocyte maintenance defects
Gene Aliases
CG9591, Dmel\CG9591, Dmel_CG9591, IntS5, Omd, intS5
Chromosome Location
Chr.3R: 13679440 - 13687210 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038168
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275613.1 NP_001262542.1 1 - 2 165 53
NM_001275614.1 NP_001262543.1 1 - 2 165 53
NM_142046.2 NP_650303.1 2 - 3 329 53
GenBank mRNA BT011415.1 - 2 - 3 329 53

View Full Product Details