Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
flfl
Entrez Gene ID
41675
Gene Name
falafel
Gene Aliases
CG9351, Dmel\CG9351, Dmel_CG9351, Flfl, PP4R3, Ppp4R3, l(3)L4179
Chromosome Location
Chr.3R: 13683921 - 13693905 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024555
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_142047.4 NP_650304.2 9 - 10 3347 66
NM_169548.3 NP_731849.2 9 - 10 3368 66
NM_169549.2 NP_731850.2 10 - 11 3468 66
GenBank mRNA AY118508.1 - 9 - 10 3309 66

View Full Product Details