Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btsz
Entrez Gene ID
3346167
Gene Name
bitesize
Gene Aliases
BcDNA:GH06647, Btsz, CG14858, CG17304, CG18280, CG31306, CG33555, CG44012, CG7343, Dmel\CG44012, Dmel_CG44012, Granulophilin, l(3)10418
Chromosome Location
Chr.3R: 14804092 - 14875488 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266756
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014622.3 NP_001014622.3 13 - 14 5039 55
NM_001014623.3 NP_001014623.1 6 - 7 4814 55
NM_001014624.3 NP_001014624.1 6 - 7 8019 55
NM_001014625.3 NP_001014625.1 11 - 12 4133 55
NM_001104324.4 NP_001097794.1 7 - 8 2245 55
NM_001144583.3 NP_001138055.1 7 - 8 1921 55
NM_001144584.3 NP_001138056.1 11 - 12 4238 55
NM_001260180.2 NP_001247109.1 13 - 14 11606 55
NM_001275653.1 NP_001262582.1 13 - 14 5066 55
GenBank mRNA AY229969.1 - 11 - 12 4117 55
AY229970.1 - 6 - 7 7811 55

View Full Product Details