Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rrp6
Entrez Gene ID
41798
Gene Name
CG7292-PB
Gene Aliases
CG7292, DmRrp6, Dmel\CG7292, Dmel_CG7292, RRP6, dRrp6, rrp6
Chromosome Location
Chr.3R: 14882498 - 14886246 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038269
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104325.2 NP_001097795.1 2 - 3 355 58
NM_001300401.1 NP_001287330.1 2 - 3 600 58

View Full Product Details