Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mf
- Entrez Gene ID
-
41824
- Gene Name
- Myofilin
- Gene Aliases
-
BcDNA:GH23305, CG6803, DmMf, Dmel\CG6803, Dmel_CG6803, Zeelin1, z1
- Chromosome Location
-
Chr.3R: 15170625 - 15181510 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038294
View Full Product Details