Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tefu
Entrez Gene ID
41839
Gene Name
telomere fusion
Gene Aliases
ATM, Atm, CG6535, Dmel\CG6535, Dmel_CG6535, Tefu, atm, atm/tefu, dATM, tef, tefu/atm
Chromosome Location
Chr.3R: 15232212 - 15242236 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0045035
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043247.2 NP_001036712.1 15 - 16 3729 87

View Full Product Details