Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Irc
Entrez Gene ID
42049
Gene Name
Immune-regulated catalase
Gene Aliases
CG8913, Dmel\CG8913, Dmel_CG8913, IRC, irc
Chromosome Location
Chr.3R: 17002875 - 17007208 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038465
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275724.1 NP_001262653.1 10 - 11 2267 75
NM_142327.3 NP_650584.1 10 - 11 2267 75
GenBank mRNA AY051443.1 - 10 - 11 2245 75

View Full Product Details