Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GckIII
- Entrez Gene ID
-
42064
- Gene Name
- Germinal centre kinase III
- Gene Aliases
-
CG5169, Dmel\CG5169, Dmel_CG5169, GCKIII, Gck III, STLK3, dGCKIII, dGckIII, wheezy
- Chromosome Location
-
Chr.3R: 17085513 - 17087988 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0266465
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142339.3 |
NP_650596.1 |
1 - 1 |
2339 |
57 |
GenBank mRNA |
BT133480.1 |
- |
1 - 1 |
2354 |
57 |
View Full Product Details