Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- beat-IIb
- Entrez Gene ID
-
42082
- Gene Name
- beaten path IIb
- Gene Aliases
-
Beat-IIb, CG14337, CG4135, CT13620, Dmel\CG4135, Dmel_CG4135, beat IIb
- Chromosome Location
-
Chr.3R: 17232253 - 17265049 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038494
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142356.3 |
NP_650613.2 |
4 - 5 |
1103 |
79 |
View Full Product Details