Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Pxt
- Entrez Gene ID
-
42131
- Gene Name
- Peroxinectin-like
- Gene Aliases
-
CG7660, Dmel\CG7660, Dmel_CG7660, Dpxt, anon-WO0172774.128, pxd, pxt
- Chromosome Location
-
Chr.3R: 17719454 - 17722955 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261987
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142391.5 |
NP_650648.3 |
2 - 3 |
1069 |
91 |
GenBank mRNA |
AY119616.1 |
- |
2 - 3 |
1009 |
91 |
View Full Product Details