Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rim
Entrez Gene ID
42150
Gene Name
Rab3 interacting molecule
Gene Aliases
CG33547, CG7301, CG7305, CG7321, DRIM, DmRIM, Dmel\CG33547, Dmel_CG33547, RIM, UNC-10/RIM, dm-Rim, rim, rim-1
Chromosome Location
Chr.3R: 17872748 - 17916578 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053547
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014630.4 NP_001014630.3 6 - 7 2366 78
NM_001260231.1 NP_001247160.1 6 - 7 2366 78
NM_001260232.2 NP_001247161.2 6 - 7 2366 78
NM_001260233.2 NP_001247162.2 6 - 7 2366 78
NM_001260234.1 NP_001247163.1 6 - 7 2366 78
NM_001260235.2 NP_001247164.2 6 - 7 2366 78
NM_001260237.2 NP_001247166.2 6 - 7 2366 78
NM_001260238.2 NP_001247167.2 6 - 7 2366 78
NM_001260239.1 NP_001247168.1 6 - 7 2366 78
NM_001260240.2 NP_001247169.2 6 - 7 2366 78
NM_001260241.2 NP_001247170.2 6 - 7 2366 78
NM_001260242.2 NP_001247171.2 6 - 7 2366 78
NM_001260243.2 NP_001247172.2 6 - 7 2366 78
NM_001275758.1 NP_001262687.1 6 - 7 2366 78

View Full Product Details